missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H7N7 Hemagglutinin Antibody (19), Biotin , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89790B
This item is not returnable.
View return policy
Description
Influenza A H7N7 Hemagglutinin Monoclonal antibody specifically detects Influenza A H7N7 Hemagglutinin in Virus samples. It is validated for ELISA
Specifications
| Influenza A H7N7 Hemagglutinin | |
| Monoclonal | |
| Biotin | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA | |
| 19 | |
| PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A Hemagglutinin H7N7 protein. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction