missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H7N7 Neuraminidase Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14650CL1
This item is not returnable.
View return policy
Description
Influenza A H7N7 Neuraminidase Polyclonal antibody specifically detects Influenza A H7N7 Neuraminidase in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Influenza A H7N7 Neuraminidase | |
| Polyclonal | |
| PBS | |
| Produced in rabbits immunized with purified, recombinant Influenza A H7N7 Neuraminidase (Accession#: AAR11367.1; His36-Ser471). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction