missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H9N2 HA (A/Hong Kong/1073/99) Monoclonal Antibody (13)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530012
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin/HA.
Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
Specifications
| Influenza A H9N2 HA (A/Hong Kong/1073/99) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody |
| ELISA | |
| 13 | |
| Unconjugated | |
| flu hemagglutinin; H9N2 HA; H9N2 hemagglutinin; Influenza H9N2; influenza hemagglutinin | |
| Recombinant Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin/HA Protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction