missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A Hemagglutinin H1 Antibody (InA139), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73160AF350
This item is not returnable.
View return policy
Description
Influenza A Hemagglutinin H1 Monoclonal antibody specifically detects Influenza A Hemagglutinin H1 in Virus samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| Influenza A Hemagglutinin H1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| InA139 | |
| Western Blot, ELISA, Sandwich ELISA | |
| HA, Hemagglutinin, Hemagglutinin HA | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/NewCaledonia/20/99 (strain H1N1). | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction