missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A M1 Antibody (1G1A12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50480DL594
This item is not returnable.
View return policy
Description
Influenza A M1 Monoclonal antibody specifically detects Influenza A M1 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Functional Assay
Specifications
| Influenza A M1 | |
| Monoclonal | |
| DyLight 594 | |
| Influenza M2 | |
| GST-Influenza A M1 fusion protein | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunofluorescence, Functional Assay | |
| 1G1A12 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction