missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A NP Recombinant Rabbit Monoclonal Antibody (014), Biotin, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 369.00 - € 850.00
Specifications
| Antigen | Influenza A NP |
|---|---|
| Clone | 014 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019290
![]() |
Invitrogen™
MA552354 |
100 μg |
€ 850.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019866
|
Invitrogen™
MA552355 |
20 μg |
€ 369.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza A NP Recombinant Monoclonal antibody specifically detects Influenza A NP in Virus samples. It is validated for ELISASpecifications
| Influenza A NP | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| Recombinant Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein/NP Protein (Met1-Asn498). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 014 | |
| ELISA | |
| Biotin | |
| Rabbit | |
| Virus | |
| Influenza nucleoprotein | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title