missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A nucleoprotein Antibody (010), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12741MFV610
This item is not returnable.
View return policy
Description
Influenza A nucleoprotein Monoclonal antibody specifically detects Influenza A nucleoprotein in Influenza virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza A nucleoprotein | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Influenza virus | |
| Purified |
| Western Blot, ELISA | |
| 010 | |
| Western Blot, ELISA | |
| Influenza A virus NP, NP, NP-A, Nucleocapsid protein, Nucleoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H3N2 (Uniprot#: P22435.2; Met1-Asn498). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction