missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A Virus H1N1 Hemagglutinin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
€ 248.00 - € 572.00
Specifications
| Antigen | Hemagglutinin/HA Peptide |
|---|---|
| Clone | 2908B |
| Dilution | 1 μg/mL (Western Blot), 3 to 15 μg/mL (Immunocytochemistry) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795855
![]() |
R&D Systems
MAB11396-100 |
100 μg |
€ 572.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746115
![]() |
R&D Systems
MAB11396-SP |
25 μg |
€ 248.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hemagglutinin/HA Peptide Monoclonal antibody specifically detects Hemagglutinin/HA Peptide in Influenza virusN1 samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
| Hemagglutinin/HA Peptide | |
| 1 μg/mL (Western Blot), 3 to 15 μg/mL (Immunocytochemistry) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell, HEK293-derived influenza a virus H1N1 Hemagglutinin with a C-terminal 6-His tag, Met1-Gln529, Accession # YP_009118626 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2908B | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Influenza virus N1 | |
| YP_009118626 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title