missing translation for 'onlineSavingsMsg'
Learn More

Influenza B Antibody (1131 (2B2)), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18082026 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18082026 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18082026 Supplier Novus Biologicals Supplier No. NB10065034JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Influenza B Hemagglutinin 2 Monoclonal antibody specifically detects Influenza B Hemagglutinin 2 in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B Hemagglutinin 2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1131 (2B2)
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Influenza B virus preparation
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-65034 exhibits nuclear staining in immunofluoresence and is therefore thought to recognize a nuclear protein. Influenza virus type B is a genus in the virus family Orthomyxoviridae responsible for acute respiratory illnesses in humans. It varies antigenically from influenza virus types A and C and is not divided into any subtypes. The gradual evolution of Influenza virus type B occurs via a series of mutations known as antigenic drift, in the surface glycoprotein haemagglutinin (HA), enabling the virus to evade the immune system and potentially re-infect the same host. HA glycoproteins from the Influenza virus type B are included in the annually updated trivalent flu vaccine, along with the Influenza virus type A subtypes H3N2 and H1N1.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.