missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B Hemagglutinin 2 Antibody (InB18), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73163AF647
This item is not returnable.
View return policy
Description
Influenza B Hemagglutinin 2 Monoclonal antibody specifically detects Influenza B Hemagglutinin 2 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Influenza B Hemagglutinin 2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza B virus. | |
| 0.2 mL | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| InB18 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Influenza B haemagglutinin 2 (HA2) | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction