Learn More
Influenza B Hemagglutinin Antibody (10A6E9C7) - (B/Phuket/3073/2013), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06570-100ul
 View more versions of this product
Product Code. 18695527
  • € 459.00 / 100µL
Description
Influenza B Hemagglutinin Monoclonal antibody specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for ELISA, Sandwich ELISA Capture
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza B Hemagglutinin
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
Virus
Purified
ELISA, Sandwich ELISA Capture
10A6E9C7
ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000
B1AG46
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza B Hemagglutinin (Accession#: ACA33493.1; Met 1-Ala 555).
100 μg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b