missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B Hemagglutinin Antibody (10H6C8G4), Novus Biologicals™
Mouse Monoclonal Antibody
€ 252.00 - € 448.00
Specifications
| Antigen | Influenza B Hemagglutinin |
|---|---|
| Clone | 10H6C8G4 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640498
|
Novus Biologicals
NBP3-06569-100ul |
100 μg |
€ 448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658896
|
Novus Biologicals
NBP3-06569-50ul |
50 μg |
€ 252.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza B Hemagglutinin Monoclonal antibody specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
| Influenza B Hemagglutinin | |
| Western Blot 1:500-1:2000, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza B Hemagglutinin (Accession#: ACA33493.1; Met 1-Ala 555). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 10H6C8G4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| B1AG46 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title