missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza B NP Monoclonal Antibody (F210J)

Description
The MA1-7309 antibody reacts with Influenza B virus NP in viral and infected tissue samples. MA1-7309 has been successfully used in Immunofluorescence applications. The MA1-7309 antibody was raised against the Influenza B Nucleoprotein.
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
Specifications
Specifications
| Antigen | Influenza B NP |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | F210J |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Influenza B; Influenza NP |
| Host Species | Mouse |
| Immunogen | Influenza B Nucleoprotein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?