missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B nucleoprotein Antibody (016), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12755B
This item is not returnable.
View return policy
Description
Influenza B nucleoprotein Monoclonal antibody specifically detects Influenza B nucleoprotein in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza B nucleoprotein | |
| Monoclonal | |
| Biotin | |
| Influenza B Virus NP, NP, Nucleoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Influenza B (Accession#: ACF54251.1; Met1-Tyr560). | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 016 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction