missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza H1N1 NA Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
H1N1 Neuraminidase Synthetic Peptide, PEP-0832, from Invitrogen.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | Influenza H1N1 NA |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Lyophilized | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction