missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING2 Antibody (AT39E5), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61159IR
This item is not returnable.
View return policy
Description
ING2 Monoclonal antibody specifically detects ING2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ING2 | |
| Monoclonal | |
| DyLight 755 | |
| ING1Linhibitor of growth family, member 1-like, ING1Lp, Inhibitor of growth 1-like protein, inhibitor of growth family, member 2, inhibitor of growth protein 2, p33ING2p32 | |
| Recombinant human ING2 (1-280aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, DNA Repair | |
| 3622 | |
| Store at 4°C in the dark. | |
| IgG1b κ |
| Western Blot, ELISA, Immunofluorescence | |
| AT39E5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction