missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING3 Goat anti-Human, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-78100-100UG
This item is not returnable.
View return policy
Description
ING3 Polyclonal specifically detects ING3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ING3 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA 1:10000 - 1:40000 | |
| NP_061944 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| 54556 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| FLJ20089, ING2, inhibitor of growth family, member 3, inhibitor of growth protein 3, MEAF4, P47, p47ING3Eaf4 | |
| This affinity purified antibody was prepared from whole Goat serum produced by repeated immunizations with a synthetic peptide corresponding aa 294-304 of Human ING3 protein (Inhibitor of growth family, member 3). This sequence only shows homology to isof | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction