missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING4 Antibody, DyLight 488, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1682G
This item is not returnable.
View return policy
Description
ING4 Polyclonal specifically detects ING4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ING4 | |
| Polyclonal | |
| Western Blot, ELISA | |
| candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein | |
| This affinity purified ING4 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 150-175 of Human ING4 protein (Inhibitor of growth family, member 4). This sequence only shows homology to isoform 1 for ING4. (Uniprot: Q9UNL4) | |
| 0.1 mL | |
| Chromatin Research, Tumor Suppressors | |
| 51147 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction