missing translation for 'onlineSavingsMsg'
Learn More

ING5 Antibody (06), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-06215-50ul

 View more versions of this product

Product Code. 18607188

  • € 220.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ING5 Monoclonal antibody specifically detects ING5 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ING5
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
ELISA
06
ELISA 1:1000-1:2000
inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ING5 (Uniprot#: Q8WYH8-2; Glu36-Gln84).
50 μg
Cancer, Tumor Suppressors
84289
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.