missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING5 Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1681H
This item is not returnable.
View return policy
Description
ING5 Polyclonal antibody specifically detects ING5 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ING5 | |
| Polyclonal | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
| This affinity purified ING5 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 125-150 of Human p28 ING5 protein (Inhibitor of growth family, member 5). (Uniprot: Q8WYH8) | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 84289 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction