missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INHBE Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121823
This item is not returnable.
View return policy
Description
INHBE Polyclonal Antibody for Western Blot, IHC (P)
Inhibins and activins inhibit and activate, respectively, the secretion of follitropin by the pituitary gland. Inhibins/activins are involved in regulating a number of diverse functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth, depending on their subunit composition. Inhibins appear to oppose the functions of activins.
Specifications
| INHBE | |
| Polyclonal | |
| Unconjugated | |
| INHBE | |
| activin beta E; activin betaE; activin beta-E chain; INHBE; inhibin beta E; inhibin beta E chain; inhibin beta E subunit; inhibin, beta E | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 16326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O08717 | |
| INHBE | |
| Recombinant protein Inhibin Beta E. The antigen corresponds to amino acid range 102-344 of the target protein. | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction