missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inhibin alpha Antibody (R1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08781R
This item is not returnable.
View return policy
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Inhibin alpha | |
| Monoclonal | |
| DyLight 550 | |
| A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
| A synthetic peptide corresponding to Human Inhibin alpha aa 233-264. Note that this peptide corresponds to the N-terminus of the mature region of the human inhibin alpha subunit. (Uniprot: P05111 ) | |
| 100 μg | |
| Apoptosis, Cancer | |
| 3623 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| R1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction