Learn More
Trajan Scientific and Medical™ Injector starter kit Siltite™ Fingertite Atas Optic suitable for GC capillary column 0.1mm to 0.25 mm I.D. SGE
Product Code. 13159025
Change view
Click to view available options
Diameter (Metric) Inner:
0.4 mm
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
13159025 0.4 mm 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13159025 Supplier Trajan Scientific and Medical™ Supplier No. 073651
Only null left
Add to basket
Add to basket
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
For Use With (Equipment) 0.1 to 0.25mm ID capillary columns
Diameter (Metric) Inner 0.4 mm
Includes 5 nuts, 10 ferrules
Description ATAS Optic Injector Starter Kit