missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INMT Polyclonal Antibody
GREENER_CHOICE

Product Code. 17269083 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17269083 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17269083 Supplier Invitrogen™ Supplier No. PA5116585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: Mouse liver, Mouse lung Immunogen sequence: MKGGFTGGDE YQKHFLPRDY LATYYSFDGS PSPEAEMLKF NLECLHKTFG PGGLQGDTLI DIGSGPTIYQ VLAACDSFQD ITLSDFTDRN REELEKWLKK EPGAYDWTPA VKFACELEGN SGRWEEKEEK LRAAVKRVLK CDVHLGNPLA PAVLPLADCV LTLLAMECAC CSLDAYRAAL CNLASLLKPG GHLVTTVTLR LPSYMVGKRE FSCVALEKEE VEQAVLDAGF DIEQLLHSPQ SYSVTNAANN GVCFIVARKK PGP.

N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INMT
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene INMT
Gene Accession No. O95050, P40936
Gene Alias Amine N-methyltransferase; aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; indolamine N-methyltransferase; Indolethylamine N-methyltransferase; Inmt; nicotine N-methyltransferase; Temt; Thioether S-methyltransferase
Gene Symbols INMT
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11185, 21743
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.