missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INMT Recombinant Rabbit Monoclonal Antibody (23GB2155), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | INMT |
|---|---|
| Clone | 23GB2155 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089327
![]() |
Invitrogen™
MA552476 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087722
|
Invitrogen™
MA552477 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INMT Recombinant Monoclonal antibody specifically detects INMT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| INMT | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O95050, P40936 | |
| 11185, 21743, 368066 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2155 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Amine N-methyltransferase; aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; indolamine N-methyltransferase; Indolethylamine N-methyltransferase; Inmt; nicotine N-methyltransferase; Temt; Thioether S-methyltransferase | |
| A synthesized peptide derived from human TEMT (1-150AA). | |
| Primary | |
| -20°C | |
| INMT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title