missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INO80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565301
This item is not returnable.
View return policy
Description
Immunogen sequence: CTELAKPLYLQ YLERALRLDH FLRQTSAIFN RNISSDDSED GLDDSNPLLP QSGDPLIQVK EEPPNSLLGE TSGAGSSGML NTYSLNGVLQ S Highest antigen sequence identity to the following orthologs - mouse 93%, rat 95%.
DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
Specifications
| INO80 | |
| Polyclonal | |
| Unconjugated | |
| Ino80 | |
| 2310079N15Rik; 4632409L19Rik; Chromatin-remodeling ATPase INO80; DNA helicase INO80; DNA helicase-related INO80 complex homolog 1; DNA helicase-related protein INO80; hINO80; homolog of yeast INO80; Ino80; INO80 complex homolog 1; INO80 complex subunit; INO80 complex subunit A; INO80 homolog; INO80 homolog (S. cerevisiae); INO80A; Inoc1; KIAA1259; putative DNA helicase INO80 complex homolog 1; RGD1310969; yeast INO80-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULG1 | |
| Ino80 | |
| Recombinant Human INO80. Recombinant protein control fragment (Product #RP-106009). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction