missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INO80C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106985
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
INO80C is a proposed core component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair.
Specifications
| INO80C | |
| Polyclonal | |
| Unconjugated | |
| INO80C | |
| C18orf37; hIes6; IES6; IES6 homolog; INO80 complex subunit C; INO80C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 125476 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q6PI98 | |
| INO80C | |
| Synthetic peptide directed towards the N-terminal region of Human INO80C. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction