Learn More
WTW™ inoLab™ Multi 9620 IDS Multi-Parameter Benchtop Meter
Use this benchtop meter for simultaneous measurement with two independent measuring channels. The WTW™ inoLab™ Multi 9620 IDS Multi-Parameter Benchtop Meter is wireless-ready and offers fatigue-proof reading on a brilliant colour display.
Brand: WTW™ 1FD560
Additional Details : Weight : 2.50000kg
Description
- Digital high performance multi-parameter device with two inputs for pH, ISE, conductivity and dissolved oxygen and comprehensive GLP functions
- Modern high-performance multi-parameter meter with two galvanically isolated measuring channels pH/mV, ISE (including incremental methods), conductivity and oxygen measurement
- With a large 16:9 colour graphic display under a hygienic glass shield and antibacterial keyboard for use, even in critical areas
- Use the new wireless modules together with the new IDS plug head sensors to be independent from cables and measure with ease under laboratory hoods or laminar flow benches
- The massive zinc die casting lower housing adds safety against interference and ensure a safe, stable stand on the lab bench
- Two USB interfaces to communicate with the computer, printer or USB memory stick allow the transfer of all data
- An activatable user administration expands the automatic GLP documentation by additional safety and traceability
Specifications
2 | |
Meter unit with universal power supply, stand and operating manual, software and UUSB cable | |
Multi 9620 IDS | |
Multi-Parameter Benchtop Meter |
LCD | |
Manual 500/10000 automatic | |
Universal power supply 100 to 240 V, 50/60 Hz | |
All except ORP |