missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iNOS (NOS2) Monoclonal Antibody (OTI11H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF813520
This item is not returnable.
View return policy
Description
iNOS (NOS2) Monoclonal antibody specifically detects iNOS (NOS2) in Human samples. It is validated for Western BlotSpecifications
| iNOS (NOS2) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q29980 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4277 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI11H3 | |
| Unconjugated | |
| MICB | |
| Hepatocyte NOS; hepatocytes; HEP-NOS; inducible nitric oxide synthase; inducible NO synthase; Inducible NOS; iNos; i-NOS; Inosl; MAC-NOS; macrophage NOS; nitric oxide synthase 2; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS; NOS type II; NOS, type II; Nos2; Nos-2; Nos2a; NOS-II; OTTMUSP00000000202; peptidyl-cysteine S-nitrosylase NOS2 | |
| Full length human recombinant protein of human MICB (NP_005922) produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction