missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPHP1 Recombinant Rabbit Monoclonal Antibody (6T10Z4)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA555505

Product Code. 30132381

  • € 457.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen Sequence: PDLGILFELG ISYIRNSTGE RGELSCGWVF LKLFDASGVP IPAKTYELFL NGGTPYEKGI EVDPSISRRA HGSVFYQIMT MRRQPQLLVK LRSLNRRSRN VLSLLPETLI GNMCSIHLLI FYRQILGDVL LKDRMSLQST DLISHPMLAT FPMLLEQPDV MDALRSSWAG KESTLKRSEK RDKEFLKSTF LLVYHDCVLP LLHSTRLPPF RWAEEETETA RWKVITDFLK QNQENQGALQ ALLSPDGVHE PFDLSEQTYD FLGEMRKNAV.

Juvenile nephronophthisis type 1 is caused by mutations of NPHP1, the gene encoding for nephrocystin. Nephrocystin interacts with p130Cas (BCAR1), proline-rich tyrosine kinase-2 (PTK2B or Pyk2), and tensin in embryonic kidney and testis, indicating that these proteins participate in a common signaling pathway. Nephrocystin and p130Cas interact in mammalian cells and both proteins prominently localize at or near sites of cell-cell contact in polarized Madin-Darby canine kidney epithelial cells. Expression of nephrocystin results in phosphorylation of Pyk2 on Tyrosine 402 as well as activation of downstream mitogen-activated protein kinases, such as ERK1 and ERK2. Nephrocystin contains a src-homology 3 (SH3) domain, which is highly conserved throughout evolution.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NPHP1
Recombinant Monoclonal
1 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
O15259, Q9QY53
NPHP1
Recombinant fusion protein containing a sequence corresponding to amino acids 463-732 of human NPHP1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
6T10Z4
Unconjugated
NPHP1
JBTS4; juvenile nephronophthisis 1 protein; nephrocystin 1; Nephrocystin1; nephrocystin-1; nephronophthisis 1 (juvenile); nephronophthisis 1 (juvenile) homolog; nephronophthisis 1 (juvenile) homolog (human); NPH1; NPHP1; SLSN1
Rabbit
Protein A
RUO
296136, 4867, 53885
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.