missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP4A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121822
This item is not returnable.
View return policy
Description
INPP4A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes an Mg++ independent enzyme that hydrolyzes the 4-position phosphate from the inositol ring of phosphatidylinositol 3,4-bisphosphate, inositol 1,3,4-trisphosphate, and inositol 3,4-bisphosphate. Multiple transcript variants encoding distinct isoforms have been described.
Specifications
| INPP4A | |
| Polyclonal | |
| Unconjugated | |
| INPP4A | |
| 107kDa; 4-Ptase-1; 9630012D15; D130048C09Rik; Inositol polyphosphate 4-phosphatase type I; Inositol polyphosphate 4-phosphatase-1; Inositol polyphosphate-4-phosphatase type I A; inositol polyphosphate-4-phosphatase, type 1; inositol polyphosphate-4-phosphatase, type I; inositol polyphosphate-4-phosphatase, type I, 107kD; inositol polyphosphate-4-phosphatase, type I, 107kDa; INPP4; Inpp4a; R74740; TVAS1; Type I inositol 3,4-bisphosphate 4-phosphatase; type I inositol-3; type I inositol-3,4-bisphosphate 4-phosphatase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3631 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96PE3 | |
| INPP4A | |
| Recombinant protein Inositol Polyphosphate-4-Phosphatase Type I 107kDa. The antigen corresponds to amino acid range 1-137 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction