missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF3e Antibody (SR1768), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22309-100ul
This item is not returnable.
View return policy
Description
eIF3e Monoclonal antibody specifically detects eIF3e in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| eIF3e | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3646 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| SR1768 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| eIF-3 p48, eIF3e, eIF3-p48, EIF3S6EIF3-P48, Eukaryotic translation initiation factor 3 subunit 6, eukaryotic translation initiation factor 3 subunit E, eukaryotic translation initiation factor 3, subunit 6 (48kD), eukaryotic translation initiation factor 3, subunit 6 48kDa, eukaryotic translation initiation factor 3, subunit E, INT6eIF3-p46, mammary tumor-associated protein INT6, murine mammary tumor integration site 6 (oncogene homolog), Viral integration site protein INT-6 homolog | |
| A synthesized peptide derived from human eIF3e (Uniprot #: P60228) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction