missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INPP5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | INPP5A |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230607
|
Novus Biologicals
NBP3-38035-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229453
|
Novus Biologicals
NBP3-38035-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INPP5A Polyclonal antibody specifically detects INPP5A in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| INPP5A | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| PBS (pH 7.3), 50% glycerol | |
| 3632 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| 43 kDa inositol polyphosphate 5-phophatase, CTCL tumor antigen HD-CL-02,5PTASE, DKFZp434A1721, EC 3.1.3.56, inositol polyphosphate-5-phosphatase, 40kD, inositol polyphosphate-5-phosphatase, 40kDa, inositol trisphosphate-5-phosphatase, 40kD, InsP3 5-phosphatase, MGC116947,5PTase, MGC116949, type I inositol-14,5-trisphosphate 5-phophatase, type I inositol-14,5-trisphosphate 5-phosphatase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-200 of human INPP5A (NP_005530.3).,, Sequence:, TAVLLVTANVGSLFDDPENLQKNWLREFYQVVHTHKPHFMALHCQEFGGKNYEASMSHVDKFVKELLSSDAMKEYNRARVYLDENYKSQEHFTALGSFYFLHESLKNIYQFDFKAKKYRKVAGKEIYSDTLESTPMLEKEKFPQDYFPECKWSRKGFIRTRWCIADCAFDLVNIHLFHDASNLVAWETSPS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title