missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INPP5D, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31642.100uL
This item is not returnable.
View return policy
Specifications
| INPP5D | |
| Polyclonal | |
| ELISA (1:40000) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user. | |
| inositol polyphosphate-5-phosphatase, 145kDa | |
| INPP5D | |
| A synthetic peptide corresponding to amino acids 960-1040 of human INPP5D (phospho Y1021). | |
| RUO | |
| 3635 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide). | |
| MGC104855/MGC142140/MGC142142/SHIP/SHIP1/SIP-145/hp51CN | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of INPP5D protein only when phosphorylated at Y1021. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction