Learn More
INPP5E Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-17797-20UL
Description
The INPP5E antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human INPP5E. This antibody recognizes human, rat, mouse antigen. The INPP5E antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| INPP5E | |
| 1000 μg/mL | |
| INPP5E, PPI5PIV | |
| 20 μL | |
| 56623 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse | |
| 150 kDa | |
| IgG |