missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INPP5J Polyclonal Antibody
GREENER_CHOICE

Product Code. 13414549
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13414549 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13414549 Supplier Invitrogen™ Supplier No. PA532875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Elephant, Horse, Pig (100%); Gibbon, Marmoset, Mouse, Rat, Bovine, Dog, Hamster, Panda, Rabbit (95%); Bat, Platypus (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Inositol and phosphatidylinositol phosphates are important for numerous cellular processes, including neuronal survival and signal transductions from growth factors, neurotransmitters and G protein coupled receptors. INPP5J (inositol polyphosphate-5-phosphatase J), also known as phosphatidylinositol 4,5-bisphosphate 5-phosphatase A, PIPP (proline-rich inositol polyphosphate 5-phosphatase), PIB5PA or INPP5, is a 1,006 amino acid cytoplasmic protein that assists in the conversion of inositol 1,4-bisphosphate from inositol 1,4,5-trisphosphate. Expressed in lung, stomach, kidney, brain, small intestine and heart, INPP5J localizes to membrane ruffles, where it may also participate in modulating inositol and phosphatidylinositol polyphosphate-binding proteins. Encoded by a gene located on human chromosome 22, INPP5J undergoes alternative splicing to produce three isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INPP5J
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene INPP5J
Gene Accession No. Q15735
Gene Alias Inositol polyphosphate 5-phosphatase J; inositol polyphosphate-5-phosphatase J; Inpp; INPP5; Inpp5j; phosphatidylinositol (4,5) bisphosphate 5-phosphatase, A; phosphatidylinositol 4,5-bisphosphate 5-phosphatase A; Pib5pa; Pipp; Proline-rich inositol polyphosphate 5-phosphatase
Gene Symbols INPP5J
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from internal region of human INPP5J / PIB5PA.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100058547, 100510987, 27124
Target Species Human, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.