missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INPP5J Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533858
This item is not returnable.
View return policy
Description
INPP5J Polyclonal Antibody for IHC (P)
Inositol and phosphatidylinositol phosphates are important for numerous cellular processes, including neuronal survival and signal transductions from growth factors, neurotransmitters and G protein coupled receptors. INPP5J (inositol polyphosphate-5-phosphatase J), also known as phosphatidylinositol 4,5-bisphosphate 5-phosphatase A, PIPP (proline-rich inositol polyphosphate 5-phosphatase), PIB5PA or INPP5, is a 1,006 amino acid cytoplasmic protein that assists in the conversion of inositol 1,4-bisphosphate from inositol 1,4,5-trisphosphate. Expressed in lung, stomach, kidney, brain, small intestine and heart, INPP5J localizes to membrane ruffles, where it may also participate in modulating inositol and phosphatidylinositol polyphosphate-binding proteins. Encoded by a gene located on human chromosome 22, INPP5J undergoes alternative splicing to produce three isoforms.
Specifications
| INPP5J | |
| Polyclonal | |
| Unconjugated | |
| INPP5J | |
| Inositol polyphosphate 5-phosphatase J; inositol polyphosphate-5-phosphatase J; Inpp; INPP5; Inpp5j; phosphatidylinositol (4,5) bisphosphate 5-phosphatase, A; phosphatidylinositol 4,5-bisphosphate 5-phosphatase A; Pib5pa; Pipp; Proline-rich inositol polyphosphate 5-phosphatase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100058547, 100338012, 100510987, 100761777, 170835, 171088, 27124, 486360, 514307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P59644, Q15735, Q9JMC1 | |
| INPP5J | |
| Synthetic 18 amino acid peptide from near C-terminus of human INPP5J / PIB5PA. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction