missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INS-IGF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145000
This item is not returnable.
View return policy
Description
Immunogen sequence: PVLFIHCPGA AGTAQGLEYR GRRVTTEPVW EEVDSSPQPQ GSESLPAQPP Antigen sequence identity: Mouse 46%; Rat 36%.
INS-IGF2 is a hybrid protein that is formed by the fusion of insulin and insulin-like growth factor 2 (IGF2) genes. It is primarily expressed in the liver and plays a role in regulating glucose metabolism and growth.
Specifications
| INS-IGF2 | |
| Polyclonal | |
| Unconjugated | |
| INS-IGF2 | |
| Insulin-like growth factor 2; Insulin-like growth factor II | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 723961 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| F8WCM5 | |
| INS-IGF2 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 117-166. Recombinant protein control fragment (Product #RP-110155). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction