missing translation for 'onlineSavingsMsg'
Learn More

INS Mouse anti-Human, Rat, Clone: IRDN/794, Abnova™

Mouse Monoclonal Antibody

Brand:  Abnova MAB14299.100ug

Product Code. 16092606

  • € 503.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Mouse monoclonal antibody raised against full length recombinant human INS.

  • High specificity
  • High reactivity
Specifications

Specifications

INS
Monoclonal
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.1-0.2 ug/mL) The optimal working dilution should be determined by the end user.
P01308
INS
Recombinant protein corresponding to full length human INS.
100 μg
Primary
Human, Rat
Liquid
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
IRDN/794
Mouse monoclonal antibody raised against full length recombinant human INS.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
ILPR/IRDN
Mouse
Protein A/G purification
RUO
3630
Store at 4°C.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)