missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INSC Polyclonal Antibody
GREENER_CHOICE

Product Code. 15766791 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15766791 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15766791 Supplier Invitrogen™ Supplier No. PA558921

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: CLQVENEHVL KSMKACVSET LSMLGQHFGQ LLELALTREV QALVRKIDAS DNIYTTESTT GNLFSLTQEG APLCRIIAKE GGVVALFKVC RQDSFRCLY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

In Drosophila, neuroblasts divide asymmetrically into another neuroblast at the apical side and a smaller ganglion mother cell on the basal side. Cell polarization is precisely regulated by 2 apically localized multiprotein signaling complexes that are tethered by Inscuteable, which regulates their apical localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INSC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Insc
Gene Accession No. Q1MX18
Gene Alias 3830422K02Rik; C730020D23; INSC; inscuteable homolog (Drosophila); inscuteable spindle orientation adaptor protein; minsc; Protein inscuteable homolog; RGD1311379
Gene Symbols Insc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human INSC. Recombinant protein control fragment (Product #RP-97541).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.