Learn More
Sampling Systems™ Inserts for use with Multi Thief
Enables the sample volume to be quickly and easily changed by changing the removable sample cells. Sampling Systems™ Insert for use with Multi Thief is ideal for taking multiple bulk samples from free flowing powders and granules.
Brand: Sampling Systems™ 1290W-05
Additional Details : Weight : 0.31680kg
Description
- 316 stainless steel construction
- Variable sample volumes
- Range of sampler lengths
- Suitable for free flowing powders and granules
- Multilevel Powder Sampler with variable Sample Volumes
Specifications
Sampler Insert | |
Stainless Steel (316) | |
5 mL |