missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSIG-1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-55244MFV610
This item is not returnable.
View return policy
Description
INSIG-1 Polyclonal antibody specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| INSIG-1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 | |
| A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot# O15503. | |
| 0.1 mL | |
| Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction | |
| 3638 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction