missing translation for 'onlineSavingsMsg'
Learn More

INSL3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NBP1-81223

 View more versions of this product

Product Code. 18288235

  • € 540.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

INSL3 Polyclonal specifically detects INSL3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

INSL3
Polyclonal
Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000 - 1:2500
P51460, P51460, P51460, P51460
INSL3
This antibody was developed against Recombinant Protein corresponding to amino acids:EMREKLCGHHFVRALVRVCGGPRWSTEARRPATGGDRELLQWLERRHLLHGLVADSNLTLGPGLQPLPQTSHHHRHHRAAATNPARYCCLSGCTQQDLLTLCPY
0.1 mL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
insulin-like 3, insulin-like 3 (Leydig cell), leydig insulin -like hormone, leydig insulin -like peptide, Leydig insulin-like peptide, Ley-I-L, MGC119818, MGC119819, Relaxin-like factor, relaxin-like factor b, RLFley-I-L, RLNL
Rabbit
Affinity Purified
RUO
3640
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only