missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSM1 Antibody (INSM1/7992R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21090JF646
This item is not returnable.
View return policy
Description
INSM1 Monoclonal antibody specifically detects INSM1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| INSM1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| IA1IA-1, insulinoma-associated 1, insulinoma-associated protein 1, Zinc finger protein IA-1 | |
| Recombinant fragment (around aa1-300) corresponding to the N-terminus of human INSM1 | |
| 0.1 mL | |
| Cancer | |
| 3642 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| INSM1/7992R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction