missing translation for 'onlineSavingsMsg'
Learn More

INSM1 Antibody (INSM1/8149R), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 286.00 - € 530.00

Specifications

Antigen INSM1
Clone INSM1/8149R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18616954
View Documents
Novus Biologicals
NBP3-20536-100ug
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18622744
View Documents
Novus Biologicals
NBP3-20536-20ug
20 μg
€ 286.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

INSM1 Monoclonal antibody specifically detects INSM1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

INSM1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer
10mM PBS with 0.05% BSA
3642
IgG κ
Protein A or G purified
INSM1/8149R
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
IA1IA-1, insulinoma-associated 1, insulinoma-associated protein 1, Zinc finger protein IA-1
Recombinant fragment (around aa1-300) corresponding to the N-terminus of human INSM1
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.