missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSM2 Antibody (INSM2/4291), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08530AF405
This item is not returnable.
View return policy
Description
INSM2 Monoclonal antibody specifically detects INSM2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| INSM2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| insulinoma-associated 2, Nbla106, zinc finger protein IA-6 | |
| Recombinant fragment of human INSM2 protein (around aa 432-540) (exact sequence is proprietary) (Uniprot: Q96T92 ) | |
| 100 μg | |
| Cancer | |
| 84684 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| INSM2/4291 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction