Learn More
Invitrogen™ INSR Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120502
Description
Positive test controls include: HepG2, HeLa, Mouse brain, Mouse kidney. The target is usually found in the following locations: Cell membrane, Single-pass type I membrane protein. Immunogen sequence: PTFLEIVNLL KDDLHPSFPE VSFFHSEENK APESEELEME FEDMENVPLD RSSHCQREEA GGRDGGSSLG FKRSYEEHIP YTHMNGGKKN GRILTLPRSN PS.
INSR is a receptor tyrosine kinase. Binding of insulin to the extracellular domain of INSR stimulates glucose uptake. INSR and IGF-1 receptors share major structural and functional similarity. The earliest cellular response to insulin stimulation is autophosphorylation of tyrosine in INSR. In humans, the INSR gene is located on chromosome 19. Defects in INSR are the cause of various insulin resistance syndromes and IGF-1R defects may also cause some forms of growth retardation.
Specifications
| INSR | |
| Polyclonal | |
| Unconjugated | |
| INSR | |
| 4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16337, 24954, 3643 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 2.3 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P06213, P15127, P15208 | |
| INSR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1281-1382 of human INSR (NP_0001992). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |