missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSRR/IR-related receptor Antibody (326929), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3055B
This item is not returnable.
View return policy
Description
INSRR/IR-related receptor Monoclonal antibody specifically detects INSRR/IR-related receptor in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| INSRR/IR-related receptor | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 326929 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| EC 2.7.10, EC 2.7.10.1, insulin receptor-related protein, insulin receptor-related receptor, IRRIR-related receptor | |
| Mouse myeloma cell line NS0-derived recombinant human INSRR, Leu24-Leu923, Accession # P14616 | |
| 0.1 mL | |
| Protein Kinase | |
| 3645 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction