Learn More
Novus Biologicals™ INSTA-Blot Rat Brain
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Membrane
Unit Size:
Each
Description
- Screening proteins from various human, mouse and rat tissues
- Contains rat brain lysate (30μg total protein per lane) on 4-20% Tris-Glycine mini gel
- Prepared from resolution by SDS-PAGE at 125V for 1 hour
- Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
- Stained with amido black for visualization of proteins
Recommended Storage
Store at room temperature
Specifications
Specifications
| For Use With (Application) | Screening proteins from various human, mouse and rat tissues |
| Quantity | 1 Membrane |
| Material | PVDF |