missing translation for 'onlineSavingsMsg'
Learn More

Instrument Check Standard 4 For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep CALMIX4100

Additional Details : Weight : 0.12600kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Packaging Triple-leached LDPE bottle
Quantity 125mL

 View more versions of this product

Product Code. 11901636

  • € 209.00 / 125mL


Estimated Shipment: 29-07-2021
Add to basket

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K50μg/mL
Al10μg/mL
As10μg/mL
Cu10μg/mL
Mn10μg/mL
Na10μg/mL
Ni10μg/mL
P10μg/mL
Pb10μg/mL
Sc10μg/mL
Zn10μg/mL
Ba1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7697-37-2
63.012
944
nitric acid
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
[N+](=O)(O)[O-]

Specifications

Instrument Check Standard 4
Components in 2% HNO3:
Al    •10Glc mug/mL
As    •10Glc mug/mL
Ba    •1Glc mug/mL
Cu    •10Glc mug/mL
Pb    •10Glc mug/mL
Mn    •10Glc mug/mL
Ni    •10Glc mug/mL
P    •10Glc mug/mL
K    •50Glc mug/mL
Sc    •10Glc mug/mL
Na    •10Glc mug/mL
Zn    •10Glc mug/mL
7697-37-2
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
Environmental
2% HNO3
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
HNO3
[N+](=O)(O)[O-]
63.012
CHEBI:48107
Triple-leached LDPE bottle
125mL
Videos
SDS
Documents
Certificates

Product intended for laboratory use only